Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wfs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031823.1 | NP_114011.1 | 5 - 6 | 735 | 63 | |
GenBank mRNA | AF136378.1 | - | 5 - 6 | 735 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |