Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032063.2 | NP_114452.1 | 7 - 8 | 1323 | 60 | |
GenBank mRNA | U78889.2 | - | 7 - 8 | 1323 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |