Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032070.1 | NP_114459.1 | 2 - 3 | 607 | 72 | |
GenBank mRNA | U89695.1 | - | 2 - 3 | 607 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |