Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsk3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032080.1 | NP_114469.1 | 6 - 7 | 858 | 66 | |
XM_006248373.2 | XP_006248435.1 | 6 - 7 | 1564 | 66 | ||
XM_006248374.2 | XP_006248436.1 | 6 - 7 | 1564 | 66 | ||
XM_006248375.2 | XP_006248437.1 | 6 - 7 | 1564 | 66 | ||
XM_008768745.1 | XP_008766967.1 | 6 - 7 | 1564 | 66 | ||
GenBank mRNA | FQ233111.1 | - | 1 - 2 | 126 | 66 | |
X73653.1 | - | 6 - 7 | 858 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Rattus norvegicus Probe spans exons |