Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032416.1 | NP_115792.1 | 5 - 6 | 594 | 59 | |
XM_006249385.2 | XP_006249447.1 | 5 - 6 | 658 | 59 | ||
GenBank mRNA | AF529165.2 | - | 5 - 6 | 471 | 59 | |
AY566467.1 | - | 5 - 6 | 538 | 59 | ||
AY566468.1 | - | 5 - 6 | 538 | 59 | ||
AY566469.1 | - | 5 - 6 | 538 | 59 | ||
BC062081.1 | - | 5 - 6 | 570 | 5621229 | 59 | |
X14977.1 | - | 5 - 6 | 594 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |