Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033352.1 | NP_203503.1 | 7 - 8 | 1995 | 90 | |
GenBank mRNA | AF131294.1 | - | 7 - 8 | 1995 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Rattus norvegicus Probe spans exons |