Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hpx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053318.1 | NP_445770.1 | 6 - 7 | 728 | 72 | |
GenBank mRNA | BC091137.1 | - | 6 - 7 | 719 | 7326169 | 72 |
FQ210231.1 | - | 6 - 7 | 729 | 72 | ||
FQ218971.1 | - | 6 - 7 | 721 | 72 | ||
FQ218998.1 | - | 3 - 4 | 497 | 72 | ||
FQ219255.1 | - | 1 - 2 | 177 | 72 | ||
FQ219309.1 | - | 6 - 7 | 727 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |