Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053358.1 | NP_445810.1 | 13 - 14 | 1187 | 117 | |
XM_006238532.2 | XP_006238594.1 | 14 - 15 | 1369 | 117 | ||
XM_006238533.2 | XP_006238595.1 | 14 - 15 | 1344 | 117 | ||
XM_006238534.2 | XP_006238596.1 | 14 - 15 | 1326 | 117 | ||
XM_006238535.2 | XP_006238597.1 | 14 - 15 | 1300 | 117 | ||
XM_006238536.2 | XP_006238598.1 | 13 - 14 | 1286 | 117 | ||
XM_006238537.2 | XP_006238599.1 | 13 - 14 | 1260 | 117 | ||
XM_006238538.2 | XP_006238600.1 | 13 - 14 | 1242 | 117 | ||
XM_006238539.2 | XP_006238601.1 | 11 - 12 | 1226 | 117 | ||
XM_008763902.1 | XP_008762124.1 | 14 - 15 | 3022 | 117 | ||
XM_008763903.1 | XP_008762125.1 | 15 - 16 | 2576 | 117 | ||
XM_008763904.1 | XP_008762126.1 | 14 - 15 | 2995 | 117 | ||
XM_008763905.1 | XP_008762127.1 | 14 - 15 | 2977 | 117 | ||
XM_008763906.1 | XP_008762128.1 | 14 - 15 | 2950 | 117 | ||
XM_008763907.1 | XP_008762129.1 | 13 - 14 | 2937 | 117 | ||
XM_008763908.1 | XP_008762130.1 | 13 - 14 | 2911 | 117 | ||
XM_008763909.1 | XP_008762131.1 | 13 - 14 | 2866 | 117 | ||
GenBank mRNA | AF121893.1 | - | 13 - 14 | 1187 | 117 | |
AY724490.1 | - | 14 - 15 | 968 | 117 | ||
BC107666.1 | - | 13 - 14 | 1171 | 7936459 | 117 | |
FQ230168.1 | - | 8 - 9 | 465 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |