Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hamp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053469.1 | NP_445921.1 | 2 - 3 | 174 | 100 | |
GenBank mRNA | AF344185.1 | - | 2 - 3 | 174 | 100 | |
FQ222617.1 | - | 2 - 3 | 193 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |