Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053528.1 | NP_445980.1 | 20 - 21 | 3415 | 108 | |
XM_006229410.2 | XP_006229472.1 | 21 - 22 | 3756 | 108 | ||
XM_008759561.1 | XP_008757783.1 | 21 - 22 | 3779 | 108 | ||
XM_008759562.1 | XP_008757784.1 | 21 - 22 | 3723 | 108 | ||
GenBank mRNA | AJ245646.1 | - | 20 - 21 | 3415 | 108 | |
AJ245647.1 | - | 20 - 21 | 3415 | 108 | ||
FQ228205.1 | - | 1 - 2 | 113 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |