Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eltd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022294.1 | NP_071630.1 | 1 - 2 | 256 | 68 | |
GenBank mRNA | AF192401.1 | - | 1 - 2 | 256 | 68 | |
AF192402.1 | - | 1 - 2 | 256 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Rattus norvegicus Probe spans exons |