Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022383.2 | NP_071778.2 | 5 - 6 | 556 | 86 | |
XM_006238830.2 | XP_006238892.1 | 5 - 6 | 656 | 86 | ||
XM_006238831.2 | XP_006238893.1 | 5 - 6 | 653 | 86 | ||
XM_006238832.2 | XP_006238894.1 | 5 - 6 | 634 | 86 | ||
GenBank mRNA | BC089801.1 | - | 5 - 6 | 556 | 7381845 | 86 |
L11930.1 | - | 4 - 5 | 458 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |