Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022400.1 | NP_071795.1 | 2 - 3 | 107 | 68 | |
XM_006229149.1 | XP_006229211.1 | 2 - 3 | 284 | 68 | ||
XM_008759413.1 | XP_008757635.1 | 2 - 3 | 849 | 68 | ||
GenBank mRNA | BC061790.1 | - | 2 - 3 | 103 | 5623666 | 68 |
U68417.1 | - | 2 - 3 | 107 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |