Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ech1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022594.1 | NP_072116.1 | 5 - 6 | 527 | 80 | |
GenBank mRNA | BC060565.1 | - | 5 - 6 | 533 | 80 | |
BC062226.1 | - | 5 - 6 | 533 | 6890529 | 80 | |
FQ214121.1 | - | 5 - 6 | 529 | 80 | ||
FQ230435.1 | - | 5 - 6 | 540 | 80 | ||
U08976.1 | - | 5 - 6 | 527 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |