Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022601.2 | NP_072123.1 | 2 - 3 | 394 | 61 | |
GenBank mRNA | BC087016.1 | - | 2 - 3 | 394 | 7128621 | 61 |
U91561.1 | - | 2 - 3 | 385 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |