Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vax1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022636.1 | NP_072158.1 | 2 - 3 | 426 | 57 | |
GenBank mRNA | AF113515.1 | - | 2 - 3 | 426 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Rattus norvegicus 1700 Probe spans exons |