Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030848.1 | NP_110475.1 | 4 - 5 | 553 | 65 | |
GenBank mRNA | D49955.1 | - | 4 - 5 | 553 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |