Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031023.1 | NP_112285.1 | 2 - 3 | 265 | 151 | |
GenBank mRNA | BC088129.1 | - | 2 - 3 | 268 | 7323494 | 151 |
FQ219359.1 | - | 2 - 3 | 274 | 151 | ||
M95768.1 | - | 2 - 3 | 265 | 151 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |