Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031023.1 | NP_112285.1 | 3 - 4 | 482 | 79 | |
GenBank mRNA | BC088129.1 | - | 3 - 4 | 485 | 7323494 | 79 |
FQ219359.1 | - | 3 - 4 | 491 | 79 | ||
FQ229296.1 | - | 1 - 2 | 105 | 79 | ||
M95768.1 | - | 3 - 4 | 482 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |