Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jup | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031047.2 | NP_112309.2 | 12 - 13 | 2437 | 100 | |
XM_006247429.1 | XP_006247491.1 | 12 - 13 | 2208 | 100 | ||
XM_006247430.2 | XP_006247492.1 | 12 - 13 | 2174 | 100 | ||
GenBank mRNA | BC065580.1 | - | 12 - 13 | 2437 | 5599724 | 100 |
FQ223166.1 | - | 12 - 13 | 2197 | 100 | ||
U58858.1 | - | 12 - 13 | 2189 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |