Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031534.2 | NP_113722.2 | 1 - 2 | 449 | 88 | |
XM_006234620.1 | XP_006234682.1 | 1 - 2 | 842 | 88 | ||
XM_006234621.2 | XP_006234683.1 | 1 - 2 | 920 | 88 | ||
XM_006234622.2 | XP_006234684.1 | 1 - 2 | 920 | 88 | ||
GenBank mRNA | X69716.1 | - | 1 - 2 | 449 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |