Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scnn1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031548.2 | NP_113736.1 | 5 - 6 | 1593 | 100 | |
GenBank mRNA | BC070902.1 | - | 5 - 6 | 1593 | 7100777 | 100 |
EU627515.1 | - | 5 - 6 | 1228 | 100 | ||
U54699.1 | - | 5 - 6 | 1260 | 100 | ||
U54700.1 | - | 5 - 6 | 1260 | 100 | ||
X70497.1 | - | 5 - 6 | 1306 | 100 | ||
X70521.1 | - | 5 - 6 | 1281 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |