Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031797.2 | NP_113985.2 | 7 - 8 | 690 | 78 | |
XM_006234597.2 | XP_006234659.1 | 7 - 8 | 764 | 78 | ||
XM_006234598.1 | XP_006234660.1 | 7 - 8 | 578 | 78 | ||
XM_008762039.1 | XP_008760261.1 | 6 - 7 | 483 | 78 | ||
GenBank mRNA | AF049882.1 | - | 7 - 8 | 604 | 78 | |
BC072498.1 | - | 7 - 8 | 690 | 7096826 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |