Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053589.1 | NP_446041.1 | 6 - 7 | 480 | 126 | |
XM_006234049.2 | XP_006234111.1 | 7 - 8 | 913 | 126 | ||
XM_006234050.2 | XP_006234112.1 | 7 - 8 | 565 | 126 | ||
GenBank mRNA | BC158585.1 | - | 6 - 7 | 1069 | 7446356 | 126 |
M83680.1 | - | 6 - 7 | 480 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |