Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Becn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034117.1 | NP_001029289.1 | 10 - 11 | 1236 | 93 | |
NM_053739.2 | NP_446191.1 | 10 - 11 | 1258 | 93 | ||
GenBank mRNA | AY033824.1 | - | 9 - 10 | 1034 | 93 | |
BC074011.1 | - | 10 - 11 | 1123 | 7100185 | 93 | |
FQ230174.1 | - | 1 - 2 | 476 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |