Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arfrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053980.1 | NP_446432.1 | 3 - 4 | 324 | 123 | |
XM_006235718.2 | XP_006235780.1 | 3 - 4 | 320 | 123 | ||
XM_006235720.2 | XP_006235782.1 | 3 - 4 | 277 | 123 | ||
GenBank mRNA | X78603.1 | - | 3 - 4 | 324 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |