Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabra4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080587.3 | NP_542154.3 | 1 - 2 | 176 | 121 | |
GenBank mRNA | S55933.1 | - | 1 - 2 | 176 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Detect Genomic DNA Rattus norvegicus Probe spans exons |