Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifa2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080775.1 | NP_542953.1 | 5 - 6 | 561 | 102 | |
XM_008762380.1 | XP_008760602.1 | 5 - 6 | 673 | 102 | ||
GenBank mRNA | M83210.1 | - | 5 - 6 | 561 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |