Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifa2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080775.1 | NP_542953.1 | 6 - 7 | 615 | 65 | |
XM_008762380.1 | XP_008760602.1 | 6 - 7 | 727 | 65 | ||
GenBank mRNA | M83210.1 | - | 6 - 7 | 615 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |