Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc5a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130746.1 | NP_570102.1 | 4 - 5 | 795 | 75 | |
GenBank mRNA | AF026554.1 | - | 4 - 5 | 795 | 75 | |
AF081204.2 | - | 1 - 2 | 383 | 75 | ||
AF081205.1 | - | 1 - 2 | 383 | 75 | ||
BC168145.1 | - | 2 - 3 | 657 | 8371761 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |