Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc5a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130746.1 | NP_570102.1 | 9 - 10 | 1283 | 60 | |
GenBank mRNA | AF026554.1 | - | 9 - 10 | 1283 | 60 | |
AF081204.2 | - | 6 - 7 | 871 | 60 | ||
AF081205.1 | - | 6 - 7 | 871 | 60 | ||
BC168145.1 | - | 7 - 8 | 1145 | 8371761 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |