Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map2k2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133283.1 | NP_579817.1 | 2 - 3 | 331 | 60 | |
XM_006240987.2 | XP_006241049.1 | 2 - 3 | 518 | 60 | ||
GenBank mRNA | BC126084.1 | - | 2 - 3 | 429 | 8376547 | 60 |
D14592.1 | - | 2 - 3 | 302 | 60 | ||
L14936.1 | - | 2 - 3 | 331 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Rattus norvegicus Probe spans exons |