Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chgb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012526.2 | NP_036658.2 | 2 - 3 | 362 | 73 | |
XM_006235072.2 | XP_006235134.1 | 3 - 4 | 272 | 73 | ||
GenBank mRNA | AF019974.1 | - | 2 - 3 | 162 | 73 | |
BC128743.1 | - | 2 - 3 | 181 | 8370781 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |