Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012707.2 | NP_036839.2 | 4 - 5 | 458 | 65 | |
GenBank mRNA | BP479145.1 | - | 4 - 5 | 458 | 65 | |
BP480555.1 | - | 4 - 5 | 433 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |