Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012747.2 | NP_036879.1 | 15 - 16 | 1573 | 90 | |
XM_006247257.2 | XP_006247319.1 | 15 - 16 | 1597 | 90 | ||
XM_006247258.2 | XP_006247320.1 | 15 - 16 | 1597 | 90 | ||
XM_006247259.2 | XP_006247321.1 | 15 - 16 | 1597 | 90 | ||
GenBank mRNA | BC087025.1 | - | 15 - 16 | 1573 | 7110655 | 90 |
X91810.1 | - | 14 - 15 | 1367 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Rattus norvegicus Probe spans exons |