Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Proc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012803.1 | NP_036935.1 | 7 - 8 | 733 | 69 | |
XM_006254526.2 | XP_006254588.1 | 7 - 8 | 805 | 69 | ||
XM_006254527.2 | XP_006254589.1 | 7 - 8 | 812 | 69 | ||
XM_008772018.1 | XP_008770240.1 | 7 - 8 | 875 | 69 | ||
GenBank mRNA | BC078879.1 | - | 6 - 7 | 710 | 7122006 | 69 |
X64336.1 | - | 7 - 8 | 733 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |