Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012939.1 | NP_037071.1 | 11 - 12 | 925 | 108 | |
XM_008766440.1 | XP_008764662.1 | 11 - 12 | 976 | 108 | ||
GenBank mRNA | BC085352.1 | - | 11 - 12 | 940 | 7115929 | 108 |
FQ213183.1 | - | 11 - 12 | 935 | 108 | ||
FQ219177.1 | - | 11 - 12 | 935 | 108 | ||
FQ219285.1 | - | 11 - 12 | 935 | 108 | ||
FQ219548.1 | - | 11 - 12 | 935 | 108 | ||
FQ219611.1 | - | 11 - 12 | 954 | 108 | ||
FQ220588.1 | - | 8 - 9 | 704 | 108 | ||
FQ223707.1 | - | 11 - 12 | 931 | 108 | ||
FQ228686.1 | - | 11 - 12 | 995 | 108 | ||
FQ228906.1 | - | 11 - 12 | 942 | 108 | ||
FQ228938.1 | - | 11 - 12 | 932 | 108 | ||
FQ228956.1 | - | 11 - 12 | 964 | 108 | ||
M38135.1 | - | 11 - 12 | 919 | 108 | ||
Y00708.1 | - | 11 - 12 | 925 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |