Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hcrtr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013064.1 | NP_037196.1 | 3 - 4 | 766 | 77 | |
GenBank mRNA | AF041244.1 | - | 3 - 4 | 766 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Rattus norvegicus Probe spans exons |