Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hcrtr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013064.1 | NP_037196.1 | 6 - 7 | 1308 | 86 | |
GenBank mRNA | AF041244.1 | - | 6 - 7 | 1308 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Rattus norvegicus Probe spans exons |