Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asns | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013079.2 | NP_037211.2 | 8 - 9 | 1319 | 68 | |
XM_006236089.2 | XP_006236151.1 | 8 - 9 | 1312 | 68 | ||
GenBank mRNA | BC081719.1 | - | 8 - 9 | 1319 | 7132312 | 68 |
U07201.1 | - | 8 - 9 | 1263 | 68 | ||
U07202.1 | - | 8 - 9 | 1263 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |