Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serinc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | DQ103710.1 | - | 7 - 8 | 892 | 65 | |
L20319.1 | - | 7 - 8 | 1013 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |