Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133424.2 | NP_596915.1 | 8 - 9 | 839 | 72 | |
GenBank mRNA | AF450248.1 | - | 8 - 9 | 840 | 72 | |
AY096238.1 | - | 8 - 9 | 764 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |