Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133608.2 | NP_598292.2 | 9 - 10 | 1978 | 58 | |
GenBank mRNA | AF239262.1 | - | 8 - 9 | 1812 | 58 | |
BC097285.1 | - | 9 - 10 | 1978 | 7441748 | 58 | |
BC097286.1 | - | 9 - 10 | 1920 | 7444297 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |