Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elovl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134382.1 | NP_599209.1 | 1 - 2 | 58 | 95 | |
XM_006243417.1 | XP_006243479.1 | 2 - 3 | 223 | 95 | ||
GenBank mRNA | AB071985.1 | - | 1 - 2 | 58 | 95 | |
HQ404314.1 | - | 1 - 2 | 58 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |