Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134461.1 | NP_604456.1 | 11 - 12 | 1712 | 61 | |
XM_006229729.2 | XP_006229791.2 | 11 - 12 | 1949 | 61 | ||
XM_006229730.2 | XP_006229792.1 | 11 - 12 | 1841 | 61 | ||
XM_006229731.2 | XP_006229793.1 | 11 - 12 | 2085 | 61 | ||
XM_008759673.1 | XP_008757895.1 | 10 - 11 | 1876 | 61 | ||
GenBank mRNA | AF484958.1 | - | 11 - 12 | 1712 | 61 | |
FQ223532.1 | - | 11 - 12 | 1806 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Detect Genomic DNA Rattus norvegicus Probe spans exons |