Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fhl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271201.1 | NP_001258130.1 | 4 - 5 | 558 | 63 | |
NM_145669.3 | NP_663702.1 | 4 - 5 | 625 | 63 | ||
GenBank mRNA | AY115564.1 | - | 4 - 5 | 587 | 63 | |
FM042472.1 | - | 4 - 5 | 558 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Rattus norvegicus Probe spans exons |