Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4x1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145675.1 | NP_663708.1 | 10 - 11 | 1269 | 79 | |
GenBank mRNA | AF439343.1 | - | 10 - 11 | 1269 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |