Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152242.2 | NP_689448.2 | 4 - 5 | 905 | 86 | |
XM_006255066.2 | XP_006255128.1 | 4 - 5 | 1957 | 86 | ||
XM_008772264.1 | XP_008770486.1 | 4 - 5 | 1107 | 86 | ||
XM_008772265.1 | XP_008770487.1 | 5 - 6 | 1371 | 86 | ||
XM_008772266.1 | XP_008770488.1 | 4 - 5 | 2617 | 86 | ||
XM_008772267.1 | XP_008770489.1 | 4 - 5 | 2880 | 86 | ||
XM_008772268.1 | XP_008770490.1 | 5 - 6 | 1204 | 86 | ||
XM_008772269.1 | XP_008770491.1 | 4 - 5 | 1986 | 86 | ||
XM_008772270.1 | XP_008770492.1 | 4 - 5 | 2462 | 86 | ||
GenBank mRNA | AF529886.1 | - | 4 - 5 | 906 | 86 | |
CV116515.1 | - | 2 - 3 | 361 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Rattus norvegicus Probe spans exons |