Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110151.1 | NP_001103621.1 | 10 - 11 | 1636 | 70 | |
NM_152847.2 | NP_690060.2 | 10 - 11 | 1636 | 70 | ||
GenBank mRNA | AB010245.1 | - | 10 - 11 | 1636 | 70 | |
AB051816.1 | - | 10 - 11 | 1636 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |