Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4f6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153318.1 | NP_695230.1 | 1 - 2 | 120 | 97 | |
XM_008765156.1 | XP_008763378.1 | 1 - 2 | 273 | 97 | ||
GenBank mRNA | U39208.1 | - | 1 - 2 | 120 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |